Anti-DKK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1522AEA 360 EUR
ABGEAP1522A
Anti-DKK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DKK2 (dickkopf WNT signaling pathway inhibitor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR