Anti-TLR5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1505AEA 360 EUR
ABGEAP1505A
Anti-TLR5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TLR5 (toll-like receptor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR