Anti-TTC33 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP15007AEA 350 EUR
ABGEAP15007A
Anti-TTC33 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TTC33 (tetratricopeptide repeat domain 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR