Anti-COPS8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14987AEA 348 EUR
ABGEAP14987A
Anti-COPS8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COPS8 (COP9 signalosome subunit 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR