Anti-CDK1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1497AEA 360 EUR
ABGEAP1497A
Anti-CDK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR