Anti-RAD51C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14962CEA 348 EUR
ABGEAP14962C
Anti-RAD51C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAD51C (RAD51 paralog C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR