Anti-CD3G Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1495AEA 360 EUR
ABGEAP1495A
Anti-CD3G Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD3G (CD3g molecule, gamma (CD3-TCR complex))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR