Anti-CLK3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14953AEA 350 EUR
ABGEAP14953A
Anti-CLK3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLK3 (CDC-like kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR