Anti-ADAM17 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1492AEA 360 EUR
ABGEAP1492A
Anti-ADAM17 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAM17 (ADAM metallopeptidase domain 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR