Anti-DYRK4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14918BEA 350 EUR
ABGEAP14918B
Anti-DYRK4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DYRK4 (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR