Anti-UTS2R Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14913CEA 348 EUR
ABGEAP14913C
Anti-UTS2R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UTS2R (urotensin 2 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR