Anti-RBM22 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14906CEA 350 EUR
ABGEAP14906C
Anti-RBM22 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RBM22 (RNA binding motif protein 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR