Anti-ALG11 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14881BEA 348 EUR
ABGEAP14881B
Anti-ALG11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALG11 (ALG11, alpha-1,2-mannosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR