Anti-NDN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14876AEA 350 EUR
ABGEAP14876A
Anti-NDN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NDN (necdin, melanoma antigen (MAGE) family member)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR