Anti-LIN28B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1485BEA 348 EUR
ABGEAP1485B
Anti-LIN28B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LIN28B (lin-28 homolog B (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR