Anti-PRKRIR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14857AEA 350 EUR
ABGEAP14857A
Anti-PRKRIR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRKRIR (protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR