Anti-LGALS9 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14853AEA 350 EUR
ABGEAP14853A
Anti-LGALS9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LGALS9 (lectin, galactoside-binding, soluble, 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR