Anti-FYN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14843BEA 350 EUR
ABGEAP14843B
Anti-FYN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR