Anti-FRK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14842BEA 348 EUR
ABGEAP14842B
Anti-FRK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FRK (fyn-related kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR