Anti-DDX3X Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1483BEA 360 EUR
ABGEAP1483B
Anti-DDX3X Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDX3X (DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR