Anti-PKD2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14827BEA 350 EUR
ABGEAP14827B
Anti-PKD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PKD2 (polycystic kidney disease 2 (autosomal dominant))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR