Anti-PTPRG Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14809CEA 348 EUR
ABGEAP14809C
Anti-PTPRG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPRG (protein tyrosine phosphatase, receptor type, G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR