Anti-NELFA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14802CEA 348 EUR
ABGEAP14802C
Anti-NELFA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NELFA (negative elongation factor complex member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR