Anti-HUNK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14794CEA 350 EUR
ABGEAP14794C
Anti-HUNK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HUNK (hormonally up-regulated Neu-associated kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR