Anti-CAMK2A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14780BEA 348 EUR
ABGEAP14780B
Anti-CAMK2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMK2A (calcium/calmodulin-dependent protein kinase II alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR