Anti-HSPBP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14772AEA 350 EUR
ABGEAP14772A
Anti-HSPBP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSPBP1 (HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR