Anti-SEPT5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14767AEA 348 EUR
ABGEAP14767A
Anti-SEPT5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEPT5 (septin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR