Anti-HTATIP2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14762AEA 348 EUR
ABGEAP14762A
Anti-HTATIP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HTATIP2 (HIV-1 Tat interactive protein 2, 30kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR