Anti-CRABP2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14756BEA 0 EUR
ABGEAP14756B
Anti-CRABP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR