Anti-SP100 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14754AEA 350 EUR
ABGEAP14754A
Anti-SP100 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SP100 (SP100 nuclear antigen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR