Anti-PTGES3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14751CEA 350 EUR
ABGEAP14751C
Anti-PTGES3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTGES3 (prostaglandin E synthase 3 (cytosolic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR