Anti-KCNMB1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14747CEA 348 EUR
ABGEAP14747C
Anti-KCNMB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCNMB1 (potassium large conductance calcium-activated channel, subfamily M, beta member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR