Anti-KCNA1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14746CEA 350 EUR
ABGEAP14746C
Anti-KCNA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCNA1 (potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR