Anti-POLR2B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14739CEA 350 EUR
ABGEAP14739C
Anti-POLR2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POLR2B (polymerase (RNA) II (DNA directed) polypeptide B, 140kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR