Anti-ALDH5A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1472AEA 350 EUR
ABGEAP1472A
Anti-ALDH5A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH5A1 (aldehyde dehydrogenase 5 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR