Anti-KDELR2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14724BEA 348 EUR
ABGEAP14724B
Anti-KDELR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KDELR2 (KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR