Anti-EPHB4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14714CEA 350 EUR
ABGEAP14714C
Anti-EPHB4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EPHB4 (EPH receptor B4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR