Anti-CSNK1E Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14708BEA 350 EUR
ABGEAP14708B
Anti-CSNK1E Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CSNK1E (casein kinase 1, epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR