Anti-PRKCB Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14697AEA 348 EUR
ABGEAP14697A
Anti-PRKCB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRKCB (protein kinase C, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR