Anti-TMPRSS13 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14675BEA 350 EUR
ABGEAP14675B
Anti-TMPRSS13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMPRSS13 (transmembrane protease, serine 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR