Anti-RUNX3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14667CEA 370 EUR
ABGEAP14667C
Anti-RUNX3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RUNX3 (runt-related transcription factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR