Anti-USP4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14657AEA 348 EUR
ABGEAP14657A
Anti-USP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USP4 (ubiquitin specific peptidase 4 (proto-oncogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR