Anti-TBC1D21 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14645AEA 348 EUR
ABGEAP14645A
Anti-TBC1D21 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TBC1D21 (TBC1 domain family, member 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR