Anti-NTRK3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14627AEA 350 EUR
ABGEAP14627A
Anti-NTRK3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NTRK3 (neurotrophic tyrosine kinase, receptor, type 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR