Anti-FER Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14620BEA 350 EUR
ABGEAP14620B
Anti-FER Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FER (fer (fps/fes related) tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR