Anti-DYRK1B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14617CEA 350 EUR
ABGEAP14617C
Anti-DYRK1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DYRK1B (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR