Anti-TAF6 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14599AEA 350 EUR
ABGEAP14599A
Anti-TAF6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAF6 (TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR