Anti-WNT3A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14596CEA 350 EUR
ABGEAP14596C
Anti-WNT3A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WNT3A (wingless-type MMTV integration site family, member 3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR