Anti-NR1I3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14587CEA 350 EUR
ABGEAP14587C
Anti-NR1I3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NR1I3 (nuclear receptor subfamily 1, group I, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR