Anti-HSD3B1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14585AEA 348 EUR
ABGEAP14585A
Anti-HSD3B1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR